Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004005671.3 | XP_004005720.1 | 1 - 2 | 498 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |