Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958449.1 | XP_027814250.1 | 2 - 3 | 668 | 86 | |
XM_027958450.1 | XP_027814251.1 | 2 - 3 | 612 | 86 |