Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004018943.4 | XP_004018992.1 | 2 - 3 | 607 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |