Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100753.2 | XP_011956143.1 | 2 - 3 | 380 | 71 | |
XM_012100754.2 | XP_011956144.1 | 2 - 3 | 380 | 71 | ||
XM_012100755.2 | XP_011956145.1 | 2 - 3 | 380 | 71 |