Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007996.4 | XP_004008045.1 | 1 - 2 | 162 | 120 | |
GenBank mRNA | FJ546080.1 | - | 1 - 2 | 122 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |