Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101121723 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004019389.4 | XP_004019438.2 | 17 - 18 | 2095 | 101 | |
XM_012101586.3 | XP_011956976.2 | 17 - 18 | 2086 | 101 |
Gene Symbol: LOC114110156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960063.1 | XP_027815864.1 | 4 - 5 | 537 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |