Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMR1NB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004022287.4 | XP_004022336.3 | 1 - 2 | 311 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |