Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIPPLY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963056.1 | XP_027818857.1 | 1 - 2 | 328 | 76 | |
XM_027963057.1 | XP_027818858.1 | 1 - 2 | 328 | 76 |