Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LLGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015098677.2 | XP_014954163.2 | 5 - 6 | 598 | 53 | |
XM_027974801.1 | XP_027830602.1 | 5 - 6 | 589 | 53 | ||
XM_027974802.1 | XP_027830603.1 | 5 - 6 | 589 | 53 | ||
XM_027974803.1 | XP_027830604.1 | 5 - 6 | 589 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |