Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011412.3 | XP_004011461.1 | 1 - 2 | 1509 | 99 | |
XM_027972561.1 | XP_027828362.1 | 2 - 3 | 267 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |