Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EOMES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015102203.2 | XP_014957689.2 | 3 - 4 | 1506 | 67 | |
XM_027957935.1 | XP_027813736.1 | 3 - 4 | 1506 | 67 | ||
XM_027957936.1 | XP_027813737.1 | 2 - 3 | 823 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |