Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC48A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094704.2 | XP_014950190.2 | 2 - 3 | 383 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |