Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002709.4 | XP_004002758.1 | 2 - 3 | 316 | 100 | |
XM_012185125.3 | XP_012040515.1 | 2 - 3 | 316 | 100 |