Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959057.1 | XP_027814858.1 | 3 - 4 | 672 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |