Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006719.4 | XP_004006768.1 | 3 - 4 | 489 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |