Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012096487.3 | XP_011951877.2 | 7 - 8 | 1087 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |