Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LARK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082079.1 | NP_001075548.1 | 3 - 4 | 1166 | 64 | |
GenBank mRNA | AF233063.1 | - | 3 - 4 | 1166 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |