Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008274882.2 | XP_008273104.1 | 664 | 60 | ||
GenBank mRNA | AJ309934.1 | - | 54 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus |