Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082183.1 | NP_001075652.1 | 2 - 3 | 253 | 88 | |
XM_008254838.2 | XP_008253060.1 | 2 - 3 | 332 | 88 | ||
XM_008254845.2 | XP_008253067.1 | 2 - 3 | 447 | 88 | ||
XM_008254846.2 | XP_008253068.1 | 2 - 3 | 366 | 88 | ||
XM_017340981.1 | XP_017196470.1 | 2 - 3 | 343 | 88 | ||
GenBank mRNA | AY989816.1 | - | 2 - 3 | 253 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |