Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082315.1 | NP_001075784.1 | 4 - 5 | 484 | 93 | |
XM_008248341.2 | XP_008246563.1 | 4 - 5 | 546 | 93 | ||
XM_008253164.2 | XP_008251386.1 | 3 - 4 | 446 | 93 | ||
GenBank mRNA | M95815.1 | - | 4 - 5 | 484 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |