Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPIND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082329.1 | NP_001075798.1 | 3 - 4 | 1179 | 86 | |
GenBank mRNA | S73493.1 | - | 3 - 4 | 1179 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |