Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101689.1 | NP_001095159.1 | 7 - 8 | 2386 | 153 | |
GenBank mRNA | X07995.1 | - | 7 - 8 | 2386 | 153 |
Gene Symbol: LCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712428.3 | XP_002712474.1 | 7 - 8 | 153 | ||
GenBank mRNA | Z27166.1 | - | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |