Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109823.1 | NP_001103293.1 | 2 - 3 | 79 | 76 | |
GenBank mRNA | EU074985.1 | - | 2 - 3 | 79 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |