Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001115911.4 | XP_001115911.3 | 1109 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |