Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095989.4 | XP_001095989.1 | 228 | 73 | ||
GenBank mRNA | JV583430.1 | - | 78 | 73 | ||
JV602646.1 | - | 114 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |