Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092593.4 | XP_001092593.2 | 799 | 65 | ||
XM_002805100.3 | XP_002805146.2 | 799 | 65 | |||
GenBank mRNA | JV636205.1 | - | 617 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |