Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266966.1 | NP_001253895.1 | 186 | 118 | ||
GenBank mRNA | JU322697.1 | - | 186 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |