Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEPSIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836917.1 | XP_028692750.1 | 1573 | 59 | ||
XM_028836918.1 | XP_028692751.1 | 1276 | 59 | |||
XM_028836921.1 | XP_028692754.1 | 1289 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |