Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107824.4 | XP_001107824.1 | 440 | 110 | ||
GenBank mRNA | JU322581.1 | - | 143 | 110 | ||
JV452944.1 | - | 317 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |