Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258129.1 | NP_001245058.1 | 234 | 78 | ||
XM_015121180.2 | XP_014976666.1 | 601 | 78 | |||
GenBank mRNA | JU327687.1 | - | 85 | 78 | ||
JU471196.1 | - | 234 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |