Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261749.1 | NP_001248678.1 | 916 | 88 | ||
GenBank mRNA | JU327739.1 | - | 428 | 88 | ||
JU474360.1 | - | 916 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |