Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261284.1 | NP_001248213.1 | 1257 | 82 | ||
GenBank mRNA | JU473128.1 | - | 1297 | 82 | ||
JV045592.1 | - | 1257 | 82 | |||
JV635760.1 | - | 1315 | 82 | |||
JV703473.1 | - | 1403 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |