Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GID4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193962.2 | NP_001180891.1 | 998 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |