Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194616.3 | NP_001181545.2 | 416 | 76 | ||
XM_001113929.4 | XP_001113929.1 | 550 | 76 | |||
XM_015111250.2 | XP_014966736.1 | 550 | 76 | |||
XM_015111251.2 | XP_014966737.1 | 510 | 76 | |||
XM_015111255.2 | XP_014966741.1 | 366 | 76 | |||
XM_028830565.1 | XP_028686398.1 | 511 | 76 | |||
XM_028830566.1 | XP_028686399.1 | 282 | 76 | |||
XM_028830567.1 | XP_028686400.1 | 252 | 76 | |||
GenBank mRNA | BQ807740.1 | - | 416 | 76 | ||
JU335937.1 | - | 407 | 76 | |||
JU476818.1 | - | 410 | 76 | |||
JV047944.1 | - | 430 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |