Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194003.2 | NP_001180932.2 | 197 | 66 | ||
GenBank mRNA | JU476067.1 | - | 222 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |