Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266753.2 | NP_001253682.1 | 590 | 110 | ||
XM_015131741.2 | XP_014987227.1 | 758 | 110 | |||
XM_015131742.2 | XP_014987228.1 | 727 | 110 | |||
XM_028843222.1 | XP_028699055.1 | 794 | 110 | |||
GenBank mRNA | JU334862.1 | - | 698 | 110 | ||
JU475883.1 | - | 699 | 110 | |||
JV043265.1 | - | 592 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |