Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047130.1 | NP_001040595.1 | 148 | 100 | ||
GenBank mRNA | U19847.1 | - | 148 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta Inventoried |