Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193749.1 | NP_001180678.1 | 432 | 142 | ||
GenBank mRNA | JU334140.1 | - | 269 | 142 | ||
JV046837.1 | - | 285 | 142 | |||
JV579453.1 | - | 301 | 142 | |||
JV649239.1 | - | 311 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |