Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112663.2 | XP_014968149.1 | 272 | 104 | ||
GenBank mRNA | JV660271.1 | - | 235 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta Inventoried |