Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265719.1 | NP_001252648.1 | 127 | 154 | ||
GenBank mRNA | JV528877.1 | - | 167 | 154 | ||
JV565041.1 | - | 127 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |