Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266441.1 | NP_001253370.1 | 760 | 115 | ||
XM_015138966.2 | XP_014994452.1 | 1792 | 115 | |||
GenBank mRNA | JU321714.1 | - | 760 | 115 | ||
JV704144.1 | - | 1508 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |