Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205183.2 | NP_001192112.1 | 923 | 106 | ||
XM_028832480.1 | XP_028688313.1 | 995 | 106 | |||
XM_028832481.1 | XP_028688314.1 | 1655 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |