Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM9SF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266461.1 | NP_001253390.1 | 360 | 104 | ||
XM_015143351.2 | XP_014998837.1 | 970 | 104 | |||
XM_015143352.2 | XP_014998838.1 | 985 | 104 | |||
XM_015143353.2 | XP_014998839.1 | 1007 | 104 | |||
XM_015143355.2 | XP_014998841.1 | 444 | 104 | |||
XM_028850507.1 | XP_028706340.1 | 603 | 104 | |||
XM_028850508.1 | XP_028706341.1 | 766 | 104 | |||
GenBank mRNA | JU320413.1 | - | 476 | 104 | ||
JU470755.1 | - | 649 | 104 | |||
JU470756.1 | - | 634 | 104 | |||
JU474007.1 | - | 634 | 104 | |||
JV045275.1 | - | 360 | 104 | |||
JV045276.1 | - | 360 | 104 | |||
JV452327.1 | - | 355 | 104 | |||
JV635649.1 | - | 505 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |