Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265993.1 | NP_001252922.1 | 1340 | 128 | ||
GenBank mRNA | JU334390.1 | - | 1340 | 128 | ||
JV704357.1 | - | 1341 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |