Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195714.1 | NP_001182643.1 | 190 | 133 | ||
XM_015118288.2 | XP_014973774.1 | 226 | 133 | |||
GenBank mRNA | CN648676.1 | - | 190 | 133 | ||
JU327181.1 | - | 111 | 133 | |||
JU489932.1 | - | 168 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |