Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265823.1 | NP_001252752.1 | 904 | 125 | ||
GenBank mRNA | JV600257.1 | - | 904 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |