Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127629.1 | NP_001121101.1 | 929 | 132 | ||
XM_028846778.1 | XP_028702611.1 | 1800 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |