Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082781.4 | XP_001082781.2 | 638 | 128 | ||
XM_028846207.1 | XP_028702040.1 | 315 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |