Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3HDM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193954.3 | NP_001180883.1 | 277 | 127 | ||
XM_015122274.2 | XP_014977760.1 | 312 | 127 | |||
XM_028838409.1 | XP_028694242.1 | 238 | 127 | |||
GenBank mRNA | JU473157.1 | - | 284 | 127 | ||
JV043252.1 | - | 286 | 127 | |||
JV528921.1 | - | 301 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |