Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASIC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102240.4 | XP_001102240.1 | 1161 | 110 | ||
XM_001102336.4 | XP_001102336.1 | 1161 | 110 | |||
XM_028846475.1 | XP_028702308.1 | 1245 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |